Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-C6-PEG3-C4-Cl | 1835705-55-9 | MFCD31560478 | >95.0% | 751.42 g/mol | C38H59ClN4O7S | 2 G
SDP

Supplier:  Medchemexpress LLC HY1036052G

Encompass_Preferred

(S,R,S)-AHPC-C6-PEG3-C4-Cl is a VHL E3 ligase ligand-linker conjugate used as a building block for PROTAC design and targeted protein degradation research. It combines a stereodefined AHPC VHL ligand with a mixed C6-PEG3-C4 linker and has been used in cell-based assays to induce degradation of HaloTag fusion proteins.

  • Used as a VHL E3 ligase ligand building block for PROTAC synthesis.
  • Soluble in DMSO at 100 mg/mL (may require ultrasonication).
  • Suitable for in vivo formulations at ≥2.5 mg/mL in recommended cosolvent systems.
  • Stable in pure form when stored at -20°C for extended periods.
  • Molecular weight 751.42 g/mol and molecular formula C38H59ClN4O7S.

Catalog No. 50-002-47672


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.