Learn More
Medchemexpress LLC (S,R,S)-AHPC-CO-C9-acid | 2172819-78-0 | 95.0% | 628.82 | 100 MG

Supplier: Medchemexpress LLC HY139345100MG
(S,R,S)-AHPC-CO-C9-acid is an E3 ligase ligand-linker conjugate that can be connected to the ligand for protein to form PROTACs. This product is for research use only and has not been fully validated for medical applications.
- E3 ligase ligand-linker conjugate
- Can be connected to ligand for protein to form PROTACs
- White to off-white solid appearance
- Target: E3 ligase ligand-linker conjugates
- Pathway: PROTAC
- Antiproliferative activity against human NCI-H69 cells
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.