Learn More
Medchemexpress LLC (S,R,S)-AHPC-isobutyl acetate-methanesulfonothioate-Me-C10-NH2 TFA | 00-00-0 | 98.9% | 924.12 g/mol | C40H60F3N5O10S3 | 5 MG

Supplier: Medchemexpress LLC HY138553A5MG
(S,R,S)-AHPC-isobutyl acetate-methanesulfonothioate Me-C10-NH2 TFA is a synthetic small-molecule intermediate provided as a trifluoroacetate salt for laboratory research use, described as part of a PROTAC BRD4 degrader. It is intended for scientific research purposes only.
- High purity suitable for research (98.86%).
- Molecular weight 924.12 g/mol.
- Chemical formula C40H60F3N5O10S3.
- Available in small milligram quantities, including 5 mg packages.
- Manufacturer provides product data sheet, SDS, and COA.
- Supplied for use in synthesis and biochemical research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.