Learn More
Medchemexpress LLC (S,R,S)-AHPC-Me-C7 ester | 2639528-48-4 | 95.5% | 50 MG

Supplier: Medchemexpress LLC HY13064050MG
(S,R,S)-AHPC-Me-C7 ester is an E3 ligase ligand-linker conjugate utilized for the synthesis of BCL-XL PROTAC degraders. It is intended for research use only and is not sold to patients. The product is a solid, appearing white to off-white, with a molecular weight of 614.80 and a chemical formula of C32H46N4O6S.
- E3 ligase ligand-linker conjugate
- Utilized for synthesis of BCL-XL PROTAC degraders
- Solid, white to off-white appearance
- Molecular weight of 614.80
- Chemical formula of C32H46N4O6S
- For research use only
- Shipped at room temperature (continental US)
- Store at -20°C in sealed storage, away from moisture
- In solvent, store at -80°C for up to 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.