Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Me-PEG2-dabrafenib | 2769753-69-5 | 99.9% | C53H62F3N9O8S3 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13734610MG

Encompass_Preferred

A selective small-molecule degrader that targets CDK17 and LIMK2, used as a chemical probe in preclinical research to study targeted protein degradation pathways.

  • Selective degradation of CDK17 and LIMK2.
  • High potency suitable for chemical biology studies.
  • High purity: 99.9%.
  • Available in multiple milligram-scale sizes for research use.
  • Molecular weight 1106.31 g·mol⁻¹.
  • Molecular formula C53H62F3N9O8S3.
  • Intended for research use only.

Catalog No. 50-002-58990


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.