Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-PEG1-N3 | 2101200-09-1 | 98.8% | 557.67 g/mol | C26H35N7O5S | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10360050MG

Encompass_Preferred

(S,R,S)-AHPC-PEG1-N3 is a VHL E3 ligase ligand-linker conjugate that incorporates an (S,R,S)-AHPC VHL ligand and a single-unit polyethylene glycol (PEG1) linker terminated with an azide. It is designed as a building block for PROTAC assembly and for click-chemistry conjugations (CuAAC), enabling attachment of functional moieties to VHL ligands.

  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Single-unit PEG linker provides improved solubility and controlled spacing between ligands.
  • High purity (>98.7%) and molecular weight 557.67 g/mol for consistent characterization.
  • Soluble in DMSO (100 mg/mL); insoluble in water (<0.1 mg/mL).
  • Stable as a powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-44905


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.