Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG1-N3 | 2101200-09-1 | MFCD31807301 | 98.8% | 557.67 g/mol | C26H35N7O5S | 2 G
SDP
Supplier:  Medchemexpress LLC HY1036002G
Encompass_Preferred
(S,R,S)-AHPC-PEG1-N3 is an azide-functionalized von Hippel-Lindau (VHL) ligand-linker conjugate used as a click chemistry reagent to attach VHL-binding modules to other molecules in PROTAC design and chemical biology studies. It contains a PEG1 spacer and a terminal azide for copper-catalyzed azide-alkyne cycloaddition, and is supplied as a solid with defined purity and molecular weight.
  • Azide terminal for copper-catalyzed click chemistry.
  • Peg1 spacer provides a short, flexible linker.
  • VHL ligand enables E3 ligase recruitment in conjugates.
  • High purity and characterized molecular weight for reproducible coupling.
  • Solid, white to light yellow, with recommended cold storage for stability.
  • Available in gram-scale pack sizes for synthesis and optimization workflows.
Catalog No. 50-002-45013