Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG1-N3 (VH032-PEG1-N3) | 2101200-09-1 | 98.8% | 557.67 g/mol | C26H35N7O5S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY103600500MG
Encompass_Preferred
(S,R,S)-AHPC-PEG1-N3 is an E3 ligase VHL ligand-linker conjugate containing an AHPC-based VHL ligand and a single-unit PEG linker terminated with an azide. It is supplied as a powder for research use and is used as a building block in PROTAC synthesis and click-chemistry conjugation.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates VHL ligand based on (S,R,S)-AHPC.
  • Features a 1-unit PEG linker for modular conjugation.
  • Azide-terminated linker enables click chemistry reactions.
  • High solubility in DMSO, insoluble in water.
  • Powder stable at -20°C for extended storage.
Catalog No. 50-002-45151