Learn More
Medchemexpress LLC (S,R,S)-ahpc-peg2-acid | 2172820-09-4 | 98.9% | C30H42N4O8S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14101610MG
Encompass_Preferred
(S,R,S)-AHPC-PEG2-acid is a PEG-based PROTAC linker that combines an AHPC E3 ligase ligand with a two-unit PEG spacer terminating in a carboxylic acid. Supplied as a high-purity solid research reagent, it is designed for use as a building block in the synthesis of PROTACs and related medicinal chemistry applications.
  • PEG2 spacer provides flexibility and improved solubility.
  • Terminal carboxylic acid enables standard amide or ester coupling chemistries.
  • AHPC ligand facilitates recruitment of E3 ligase for targeted degradation strategies.
  • High reported purity supports reliable analytical and synthetic work.
Catalog No. 50-002-59305