Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-acid | 2172820-09-4 | 98.9% | C30H42N4O8S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14101650MG
Encompass_Preferred
(S,R,S)-AHPC-PEG2-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. They function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets the protein of interest
  • Functions by leveraging the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-54541