Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-C4-CI | 1835705-57-1 | MFCD31560480 | ≥98.0% | 651.26 g/mol | C32H47ClN4O6S | 50 MG

Supplier: Medchemexpress LLC HY10360750MG
(S,R,S)-AHPC-PEG2-C4-Cl is a VHL ligand-linker conjugate used as a building block in PROTAC and targeted protein degradation research. It features an (S,R,S)-configured AHPC VHL ligand linked via a PEG2-C4 spacer to a terminal chloride, typically supplied as a light yellow oil and provided at high purity for chemical biology applications.
- Includes a PEG2-C4 linker with a terminal chloride for conjugation
- Contains an (S,R,S)-configured AHPC VHL ligand moiety
- High purity suitable for synthesis and biological assays (≥98.0%)
- Molecular weight 651.26 g/mol; formula C32H47ClN4O6S
- Supplied as an oil; handle and store per safety data sheet
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.