Learn More
Medchemexpress LLC HY-103599 100mg Medchemexpress, (S,R,S)-AHPC-PEG2-N3 CAS:2010159-45-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103599100MG
Encompass_Preferred
Medchemexpress, HY-103599 100mg (S,R,S)-AHPC-PEG2-N3 CAS:2010159-45-0 (S,R,S)-AHPC-PEG2-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 2-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8134