Learn More
Medchemexpress LLC (S,R,S)-Ahpc-peg2-n3 | 2010159-45-0 | 99.9% | 601.72 g/mol | C28H39N7O6S | 10 MG

Supplier: Medchemexpress LLC HY10359910MG
(S,R,S)-AHPC-PEG2-N3 is a VHL E3 ligase ligand-linker conjugate that incorporates an (S,R,S)-AHPC-based VHL ligand linked to a two-unit PEG spacer terminating in an azide group. It is supplied as a research reagent for use in PROTAC development and for click-chemistry conjugation.
- VHL E3 ligase ligand-linker conjugate.
- Azide terminal group for click chemistry conjugation.
- Two-unit PEG spacer provides a flexible linker.
- Molecular weight 601.72 g/mol.
- Soluble in DMSO (125 mg/mL) and ethanol (50 mg/mL); ultrasonic assistance recommended.
- Store at -20°C under nitrogen; in solvent, store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.