Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG2-N3 | 2010159-45-0 | 99.9% | 601.72 g/mol | C28H39N7O6S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10359950MG
Encompass_Preferred
(S,R,S)-AHPC-PEG2-N3 is an E3 ligase ligand-linker conjugate containing an (S,R,S)-AHPC VHL ligand connected to a two-unit PEG linker and terminating in an azide. It is provided as a click-chemistry reagent for PROTAC assembly and linker installation.
  • Contains a terminal azide compatible with CuAAC and SPAAC reactions.
  • Designed for conjugation to alkyne-bearing molecules and strain-promoted partners.
  • PEG2 linker provides flexibility and solubility for linker installation.
  • Supplied as a research-grade reagent with characterized molecular weight and formula.
Catalog No. 50-002-44633