Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG3-NH2 | 2010159-56-3 | MFCD31727698 | 99.3% | 619.77 | C30H45N5O7S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY103602A50MG
Encompass_Preferred
(S,R,S)-AHPC-PEG3-NH2 is a synthesized E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC VHL-binding motif with a three-unit polyethylene glycol (PEG3) linker and a terminal amine. It is provided as the free base for use in PROTAC design and E3 ligase binding studies.
  • VHL ligand-linker conjugate for PROTAC design and E3 ligase studies
  • Three-unit PEG linker provides flexibility and solubility
  • Terminal amine enables straightforward conjugation to target ligands
  • High purity (99.26%) and molecular weight 619.77 g·mol-1
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month)
  • Supplied as a 50 MG package
Catalog No. 50-002-44838