Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG4-NH2 hydrochloride | 2064292-52-8 | 98.2% | 700.29 g/mol | C32H50ClN5O8S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10360425MG
Encompass_Preferred
(S,R,S)-AHPC-PEG4-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC-based VHL ligand with a four-unit polyethylene glycol (PEG4) linker terminated in a primary amine; provided as the hydrochloride salt and supplied as a solid for research use.
  • Used as an E3 ligase recruiting module in PROTAC design.
  • PEG4 linker provides a flexible, water-soluble spacer for linker optimization.
  • Terminal amine enables straightforward conjugation to target-binding ligands.
  • High purity (~98.2%) solid suitable for synthetic and analytical workflows.
  • Chemical formula and structure information provided for synthesis planning and cheminformatics.
Catalog No. 50-002-45015