Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG5-Boc | 2923734-82-9 | 95.3% | 807.01 g/mol | C40H62N4O11S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1319595MG
Encompass_Preferred
(S,R,S)-AHPC-PEG5-Boc is an E3 ligase ligand-linker conjugate that combines an (S,R,S)-AHPC-derived VHL ligand with a PEG5 spacer and a Boc protecting group. It is designed for use in targeted protein degradation research and PROTAC synthesis, and is supplied at high purity for medicinal chemistry applications. CAS 2923734-82-9, molecular weight 807.01 g/mol, purity ≈95.3%.
  • Boc-protected VHL ligand-linker conjugate suitable for PROTAC development.
  • Peg5 linker provides flexible spacing for bifunctional molecule design.
  • High purity supports medicinal chemistry and analytical work.
  • Available in small mass quantities to support discovery research.
  • Soluble in DMSO for convenient formulation and handling.
  • Stable when stored cold under inert atmosphere for longer-term storage.
Catalog No. 50-002-62592