Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG5-Boc | 2923734-82-9 | 95.3% | C40H62N4O11S | 10MG

Supplier: Medchemexpress LLC HY13195910MG
(S,R,S)-AHPC-PEG5-Boc is an E3 ligase ligand-linker conjugate that couples an (S,R,S)-AHPC-based VHL ligand to a PEG5 linker with a Boc protecting group. Supplied as a solid research reagent, it is intended for use as the ligand-linker component in PROTACs such as Cdc20-targeting degraders.
- Incorporates the (S,R,S)-AHPC VHL ligand
- Contains a PEG5 linker with Boc protection
- Provided as a solid, light yellow to yellow material
- Molecular formula: C40H62N4O11S
- Molecular weight: 807.01 g·mol⁻¹
- Reported purity: 95.3%
- Storage: -20 °C under nitrogen; in solvent -80 °C (6 months) or -20 °C (1 month)
- Available in milligram to gram sizes for research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.