Learn More
Medchemexpress LLC (S,R,S)-AHPC-PEG5-COOH | 2172820-14-1 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130271100MG
Encompass_Preferred
(S,R,S)-AHPC-PEG5-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 5-unit PEG linker, commonly used in PROTAC technology. It functions by recruiting the VHL protein, a key substrate recognition subunit of Cullin RING E3 ubiquitin ligase complexes, to induce ubiquitination and subsequent proteasomal degradation of target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Includes 5-unit PEG linker
  • Used in PROTAC technology
  • Recruits VHL protein to induce ubiquitination
  • Promotes proteasomal degradation of target proteins
  • For research use only
Catalog No. 50-003-39571