Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride | 2376990-28-0 | 98.2% | 660.70 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136183100MG
Encompass_Preferred
(S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and an alkyl linker used for PROTAC degrader. It is intended for research use only.
  • E3 ligase ligand-linker conjugate
  • Integrates the VH032 based VHL ligand and an alkyl linker
  • Designed for use in PROTAC degrader applications
Catalog No. 50-003-50152