Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride | 2376990-28-0 | 98.2% | 660.70 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13618350MG
Encompass_Preferred
(S,R,S)-AHPC-phenol-alkylC6-amine dihydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the VH032 based VHL ligand and an alkyl linker used for PROTAC degrader.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates VH032 based VHL ligand
  • Used for PROTAC degrader
  • For research use only
Catalog No. 50-003-49070