Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride | 2376990-26-8 | 96.3% | 632.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13618450MG
Encompass_Preferred
(S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Utilizes a linker used in PROTAC technology
  • Suitable for cancer and cancer targeted therapy research
  • Functions as an E3 ligase ligand-linker conjugate inhibitor
Catalog No. 50-003-48180