Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride | 2376990-26-8 | 96.33% | 632.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136184100MG
Encompass_Preferred
(S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used in PROTAC technology. It is for research use only and not sold to patients.
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49352