Learn More
Medchemexpress LLC (S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride | 2376990-26-8 | >98.0% | C28H43Cl2N5O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13618410MG
Encompass_Preferred
(S,R,S)-AHPC-phenol-C4-NH2 dihydrochloride is a small-molecule VHL ligand derivative supplied as the dihydrochloride salt for use in PROTAC and E3 ligase recruitment research. It is reported as a solid reagent with high purity and is accompanied by analytical documentation suitable for research and development (R&D) applications.
  • Used for VHL recruitment in PROTAC design and protein degradation studies.
  • Provided as a dihydrochloride salt for improved solubility and handling.
  • High reported purity suitable for chemical biology research.
  • Supplied in small-scale pack sizes for research and development use.
  • Accompanied by analytical data such as COA and SDS for quality verification.
Catalog No. 50-002-59140