Learn More
Medchemexpress LLC HY-126456 10mg Medchemexpress, (S,R,S)-AHPC-propargyl CAS:2098799-78-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12645610MG
Encompass_Preferred
Medchemexpress, HY-126456 10mg (S,R,S)-AHPC-propargyl CAS:2098799-78-9 (S,R,S)-AHPC-propargyl (VH032-propargyl) is a VHL ligand which is used in “click reaction” for PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9728