Learn More
Medchemexpress LLC (S,R,S)-CO-C2-acid | 2172819-72-4 | MFCD32898802 | ≥98.0% | 530.64 g·mol⁻¹ | C26H34N4O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1318665MG
Encompass_Preferred
(S,R,S)-CO-C2-acid is a VH032-derived small molecule VHL ligand provided as an off-white to light-yellow solid for use in chemical biology and medicinal chemistry research, including PROTAC development. It has molecular formula C26H34N4O6S, a molecular weight of 530.64 g·mol⁻¹, and reported purity ≥98%. The compound serves as a versatile building block for conjugation and E3 ligase recruitment in degrader design.
  • High chemical purity suitable for research applications.
  • Defined stereochemistry for consistent biological performance.
  • Acts as a VHL E3 ligase ligand for PROTAC and degrader design.
  • Compatible with standard amide coupling and linker installation chemistries.
  • Supplied in small-scale quantities for discovery and synthesis workflows.
Catalog No. 50-002-62676