Learn More
Medchemexpress LLC (S)-Rivastigmine-d6 tartrate | 194930-00-2 | 99.0% | 406.46 | 10 MG

Supplier: Medchemexpress LLC HY11017AS1MG
(S)-Rivastigmine-d6 (tartrate) is a deuterium-labeled (S)-Rivastigmine, which acts as a cholinesterase inhibitor. It is intended for research use only.
- Deuterium-labeled (S)-Rivastigmine
- Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into the drug molecule
- Appearance: solid, white to off-white
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: -20°C, sealed storage, away from moisture
- Soluble in DMF (≥ 25 mg/mL), DMSO (≥ 16 mg/mL), and ethanol (≥ 16 mg/mL)
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.