Learn More
Medchemexpress LLC HY-B0392 10mg Medchemexpress, (S)-(+)-Rolipram CAS:85416-73-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB039210MG
Encompass_Preferred
Medchemexpress, HY-B0392 10mg (S)-(+)-Rolipram CAS:85416-73-5 (S)-(+)-Rolipram ((+)-Rolipram) is a cyclic AMP(cAMP)-specific phosphodiesterase (PDE) inhibitor, with an IC50 of 1100 nM. (S)-(+)-Rolipram can suppresse tumor necrosis factor-alpha (TNFα) production by human mononuclear cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2514