Learn More
AdipoGen S-Roscovitine
SDP
Supplier:  AdipoGen AGMRC0002M005
Encompass_Preferred
Chemical. CAS 186692-45-5. Formula C19H26N6O. MW 354.5. S-enantiomer of R-roscovitine Prod. No. MR-C0001. Potent selective cyclin-dependent kinase 1 CDK1 inhibitor. Potential neuroprotectant for stroke. Crosses the blood brain barrier. Inhibits CDK5, consequently blocking hypoxia-induced apoptosis in neurons. Potential anti-inflammatory compound. Potential antidiabetic compound.
Catalog No. 50-256-2164