Learn More
Selleck Chemical LLC S-Ruxolitinib (INCB018424) S2902-5mg
SDP
Supplier:  Selleck Chemical LLC S29025MG
Encompass_Preferred
S-Ruxolitinib is the chirality of INCB018424 which is the first potent selective JAK1/2 inhibitor to enter the clinic with IC50 of 3 3 nM/2 8 nM 130-fold selectivity for JAK1/2 versus JAK3 Phase 3
Catalog No. 50-311-2278