Learn More
Strem, An Ascensus Company CAS# 494227-35-9. 100mg. (S,S)-(+)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(dicyclohexylphosphino)ferrocene, min. 97%. MFCD04117702
SDP
Supplier:  Strem, An Ascensus Company 260240100MG
Encompass
CAS# 494227-35-9. 100mg. (S,S)-(+)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(dicyclohexylphosphino)ferrocene, min. 97%. MFCD04117702. Molecular Weight: 844.95. Molecular Formula: C52H74FeN2P2. Color/form: orange-red solid Strem# 26-0240. http://www.strem.com/catalog/v/26-0240/
Catalog No. 50-138-5453