Learn More
Strem, An Ascensus Company CAS# 876608-69-4. 100mg. (S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis[di(3,5-dimethyl-4-methoxyphenyl)phosphino]ferrocene, min. 97%. MFCD04117704
SDP
Supplier:  Strem, An Ascensus Company 260248100MG
Encompass
CAS# 876608-69-4. 100mg. (S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis[di(3,5-dimethyl-4-methoxyphenyl)phosphino]ferrocene, min. 97%. MFCD04117704. Molecular Weight: 1053.08. Molecular Formula: C64H74FeN2O4P2. Color/form: yellow to orange-red solid Strem# 26-0248. http://www.strem.com/catalog/v/26-0248/
Catalog No. 50-901-10962