Learn More
Strem, An Ascensus Company CAS# 850444-36-9. 100mg. (S)-(-)-[(S)-2-Diphenylphosphinoferrocenyl](N,N-dimethylamino)(2-diphenylphosphinophenyl)methane, min. 97% TANIAPHOS. MFCD04117709
SDP
Supplier:  Strem, An Ascensus Company 261155100MG
Encompass
CAS# 850444-36-9. 100mg. (S)-(-)-[(S)-2-Diphenylphosphinoferrocenyl](N,N-dimethylamino)(2-diphenylphosphinophenyl)methane, min. 97% TANIAPHOS. MFCD04117709. Molecular Weight: 687.57. Molecular Formula: C43H39FeNP2. Color/form: orange pwdr. Strem# 26-1155.
Catalog No. 50-901-11025