Learn More
Chemglass Life Sciences Chemglass Stainless Steel Adapter, M30 Male to M30 Male, Hex Style, for Huber circulator fittings
SDP
Supplier:  Chemglass Life Sciences CG1969M105
Encompass
The Chemglass stainless steel M30 adapter connects two M30 fittings to provide a secure, leak-resistant connection for circulator and temperature-control systems. The hex-style body permits wrench tightening for easy installation and reliable sealing.
  • Stainless steel construction for corrosion resistance.
  • Hex-style body for wrench tightening.
  • M30 male-to-male configuration for connecting M30 fittings.
  • Suitable for circulator and temperature-control systems.
  • Compact form factor for tight installation spaces.
Catalog No. NC2588387