Learn More
Medchemexpress LLC (S,S)-BMS-984923 | 1375752-77-4 | 98.8% | 374.8 g/mol | C22H15ClN2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141848A5MG
Encompass_Preferred
(S,S)-BMS-984923 is the (S,S)-enantiomer of an mGluR5 silent allosteric modulator provided as a research reagent. It is described as a less active enantiomer versus the parent compound and is intended for in vitro research applications as a solid powder.
Catalog No. 50-002-20976