Learn More
BOSTON FLUID SYSTEMS INC S/S IN-LINE PARTICULT FLTR,1/8
Supplier:  BOSTON FLUID SYSTEMS INC SS2F2
Encompass
Stainless Steel In-Line Particulate Filter, 1/8 in. Swagelok Tube Fitting, 2 Micron Pore Size
Catalog No. NC1882824