Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,S,R,S,R)-Boc-Dap-NE | 3026595-14-9 | 99.8% | C23H36N2O5 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY78931C100MG

Encompass_Preferred

This product, identified as (S,S,R,S,R)-Boc-Dap-NE, is categorized as a laboratory chemical for the manufacture of substances. It is intended for research use only and must be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities. The product has been tested and complies with the given specifications.

  • Appearance: White to off-white solid
  • Chemical formula: C23H36N2O5
  • Molecular weight: 420.54
  • Purity: 99.78% (HPLC)
  • Storage: 4°C under nitrogen; in solvent, -80°C for 6 months or -20°C for 1 month under nitrogen. Shipping at room temperature if less than 2 weeks.

Catalog No. 50-004-35032


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.