Learn More
Medchemexpress LLC (S,S,R,S,R)-Boc-Dap-NE | 3026595-14-9 | 99.8% | C23H36N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY78931C50MG
Encompass_Preferred
This product, (S,S,R,S,R)-Boc-Dap-NE, is a white to off-white solid with a molecular weight of 420.54, designed for laboratory research and the manufacture of substances. It is important to note that this product has not been fully validated for medical applications and is for research use only.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-34901