Learn More
Medchemexpress LLC (S,S,S)-AHPC-Boc | 1797406-74-6 | 97.2% | 530.68 g/mol | C27H38N4O5S | 100MG
SDP
Supplier:  Medchemexpress LLC HY123109A100MG
Encompass_Preferred
(S,S,S)-AHPC-Boc is the Boc-protected stereoisomer of AHPC (also referred to as VH032), supplied as a research-grade small molecule for chemical biology and medicinal chemistry applications. The compound contains a tert-butoxycarbonyl (Boc) protecting group and defined stereochemistry, making it useful as a synthetic intermediate and building block for ligand and probe assembly, including E3 ligase ligand development.
  • High purity suitable for analytical and synthetic use.
  • Boc-protected functionality enables selective deprotection strategies.
  • Defined stereochemistry for stereospecific synthesis and SAR studies.
  • Solid form that is stable under recommended storage conditions.
  • Available in small research quantities for laboratory workflows.
Catalog No. 50-002-35537