Learn More
Medchemexpress LLC (S,S,S,S,R)-Boc-Dap-NE | 3026594-75-9 | C23H36N2O5 | 50 MG
Supplier: Medchemexpress LLC HY78931B50MG
This product is identified as (S,S,S,S,R)-Boc-Dap-NE. Its relevant uses include laboratory chemicals and the manufacture of substances. Chemically, it has a molecular formula of C23H36N2O5 and a molecular weight of 420.54. The product appears as a solid and has a relative density of 1.125±0.06g/cm³. According to hazard identification, this substance is classified as acutely toxic, causing skin and serious eye irritation, and respiratory tract irritation. It carries a "Warning" signal word with hazard statements such as "Harmful if swallowed," "Causes skin irritation," "Causes serious eye irritation," and "May cause respiratory irritation."
- Identified for use as laboratory chemicals
- Suitable for the manufacture of substances
- Appears as a solid
- Relative density of 1.125±0.06g/cm³
- Classified as acutely toxic
- Causes skin and serious eye irritation
- May cause respiratory tract irritation