Learn More
Medchemexpress LLC (S)-seco-duocarmycin SA | 152785-82-5 | MFCD29127463 | 99.0% | 513.93 | C25H24ClN3O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY129356A10MG
Encompass_Preferred
(S)-Seco-Duocarmycin SA is the S-enantiomer of Seco-Duocarmycin SA, a DNA-alkylating antitumor antibiotic commonly used as a cytotoxic payload for antibody-drug conjugates (ADCs). It induces concentration-dependent apoptotic cell death and can cause cell-cycle arrest in S and G2/M phases.
  • Acts as a DNA alkylator and antitumor antibiotic.
  • Used as a cytotoxic payload for antibody-drug conjugates (ADCs).
  • Induces concentration-dependent apoptotic cell death.
  • Can cause cell-cycle arrest in S and G2/M phases.
  • Reported potency approx. 10 pM in antitumor assays.
  • Chemical identifiers: CAS 152785-82-5; formula C25H24ClN3O7; molecular weight 513.93.
Catalog No. 50-002-03941