Learn More
Apexbio Technology LLC SuperSignal ECL Chemiluminescent Substrate Detection Kit (Ultrasensitive) 500mL (250 mL each for A and B)
SDP
Supplier:  Apexbio Technology LLC K1233500
Encompass_Preferred
The SuperSignal ECL Chemiluminescent Substrate Detection Kit (Ultrasensitive) is an enhanced ECL substrate with exceptionally high sensitivity and an extended signal duration up to 16 hours It allows detection of low femtogram-level (based on HRP concentration) target proteins using HRP in immunoblotting assays demonstrating superior sensitivity prolonged luminescence duration and lower background signals Features 1) High Sensitivity Allows femtogram-level detection of protein bands on nitrocellulose or PVDF membranes under proper conditions 2) Quantitative Performance Quantifiable detection of signals span two orders of magnitude 3) Bright Signal Produces clear bright signals easily captured by film or imaging systems 4) Extended Signal Longevity Measurable chemiluminescent signal for up to 16 hours under optimized conditions 5) Stable Reagents Stable for 1 year at 4 C and 6 months at RT 6) Economical Pricing Enables effective detection even with very low Ab conc
Catalog No. 50-310-7290