Learn More
Medchemexpress LLC (S)-Subasumstat | 1858282-76-4 | 99.6% | 578.10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY111789A10MG
Encompass_Preferred
(S)-Subasumstat is the isomer of Subasumstat (HY-111789). Subasumstat (TAK-981) is a first-in-class and selective inhibitor of the SUMOylation enzymatic cascade, possessing potential immune-activating and antineoplastic activities.
  • Possesses potential immune-activating activities.
  • Exhibits antineoplastic activities.
  • Relevant for cancer research.
  • Relevant for inflammation or immune system disease research.
Catalog No. 50-004-39362