Learn More
Medchemexpress LLC (S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate | 1273577-11-9 | MFCD18633590 | ≥97.0% | 230.31 g/mol | C11H22N2O3 | 25 G

Supplier: Medchemexpress LLC 25GHYW069334
Chiral piperazine intermediate used as a building block in the synthesis of active pharmaceutical compounds.
- Chiral (S) enantiomer.
- Used as a drug intermediate for synthesis of various active compounds.
- CAS number 1273577-11-9.
- Molecular formula C11H22N2O3 and molecular weight 230.31 g/mol.
- Typically supplied in a 25 g unit.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.