Learn More
Medchemexpress LLC S-tert-butyl-L-cysteine hydrochloride | 2481-09-6 | MFCD00037426 | 98.0% | 213.73 g/mol | C7H16ClNO2S | 500 G
SDP
Supplier:  Medchemexpress LLC HYW014238500G
Encompass_Preferred
S-tert-butyl-L-cysteine hydrochloride is the hydrochloride salt of an S-tert-butyl substituted L-cysteine - an amino acid derivative used in synthetic chemistry and biochemical research. Supplied as a powder with reported purity of 98.0%, it is suitable for incorporation into peptide synthesis and studies of modified amino acids. Store powder at -20°C for long-term stability and at 4°C for shorter storage; in solution store at -80°C or -20°C depending on duration.
  • High purity (98.0%) for research use.
  • Powder form suitable for synthetic and analytical applications.
  • Useful as a modified amino acid in biochemical studies.
  • Defined storage conditions for long- and short-term stability.
  • Available in multiple pack sizes to support scale-up.
Catalog No. 50-003-03863