Learn More
Medchemexpress LLC (S)-(-)-tert-Butylsulfinamide | 343338-28-3 | MFCD05861480 | 99.9% | 121.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY4022910MM/1ML
Encompass_Preferred
(S)-(-)-tert-Butylsulfinamide is a compound containing a chiral sulfur atom. It can be used in the synthesis of other compounds, specifically as a chiral auxiliary in the asymmetric synthesis of organic compounds.
  • Contains a chiral sulfur atom
  • Can be used as a chiral auxiliary
  • Useful in the asymmetric synthesis of organic compounds
Catalog No. 50-004-05132