Learn More
Medchemexpress LLC S-tert-Leucine N-methylamide | 89226-12-0 | 98.0% | C7H16N2O | 1 G
SDP
Supplier:  Medchemexpress LLC HY792711G
Encompass_Preferred
S-tert-Leucine N-methylamide is a leucine derivative. Amino acids and amino acid derivatives, such as this product, have been commercially used as ergogenic supplements. They are recognized to be beneficial as ergogenic dietary substances, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage.
  • Leucine derivative
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • For research use only
Catalog No. 50-004-33010