Learn More
Medchemexpress LLC S-tert-Leucine N-methylamide | 89226-12-0 | 98.0% | C7H16N2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY79271100MG
Encompass_Preferred
S-tert-Leucine N-methylamide is a leucine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, affecting the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Leucine derivative
  • Used as an ergogenic supplement
  • Affects the secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-33170