Learn More
Abcam (S)-Timolol maleate, non-selective beta adrenoceptor antagonist, 100MG
Supplier:  Abcam AB120784100MG
Encompass_Preferred
MW 432.49 g/mol, Purity >99%. Potent non-selective β adrenoceptor antagonist (Ki values are 1.97 and 2.0 nM for β₁ and β₂ receptor subtypes, respectively). 50% bioavailability following oral administration. Does not cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7275