Learn More
Medchemexpress LLC (S)-TNG260 | 2935964-93-3 | C20H18FN3O2S | 1 ML

Supplier: Medchemexpress LLC HY153358A10MM/1ML
(S)-TNG260 is an isomer of TNG260, which functions as a CoREST selective deacetylase (CoreDAC) inhibitor. It demonstrates significant selectivity, inhibiting HDAC1 with 10-fold preference over HDAC3. This compound is known to induce HDAC1 inhibition, effectively reversing anti-PD1 resistance associated with STK11 deletion. Additionally, it reduces intratumoral neutrophil infiltration and exhibits immune-mediated cell killing.
- CoREST selective deacetylase (CoreDAC) inhibitor
- Inhibits HDAC1 with 10-fold selectivity over HDAC3
- Reverses anti-PD1 resistance driven by STK11 deletion
- Reduces intratumoral infiltration of neutrophils
- Exhibits immune-mediated cell killing
- Purity of 98.84%
- Molecular weight is 383.44
- Appears as an off-white to light yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.